missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FNTB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | FNTB |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16878633
|
Proteintech
11507-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16868633
|
Proteintech
11507-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ZNF24 is a new candidate transcription factor.Specifications
FNTB | |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P49356, Q02293, Q8K2I1 | |
110606, 2342, 64511 | |
FNTB Fusion Protein Ag2061 | |
Primary | |
-20°C | |
FNTB |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FNTB, FPTB, FTase beta | |
Fntb | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |