Learn More
Scientific Industries SI™ Foam Inserts for Tubes
Keeps tubes upright and immobile while in use with Vortex-Genie 2. Scientific Industries SI™ Foam Inserts for Tubes fit snugly inside a Vortex-Genie 2 Recessed Platform.
Brand:  Scientific Industries SI™ 503-0279-00
Additional Details : Weight : 0.75000kg
 View more versions of this product
Product Code. 15507325
  • 275.00 SEK / Each
Estimated Shipment: 20-05-2024
to see stock.
Add to basket
Description
Description
  • Constructed of high-density foam fitting several 9 tube sizes
  • For use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
  • One insert is included in the Large Sample Set
Specifications
Specifications
Tubes
High-density foam
8
14 to 29 mm
Vortex-Genie 2 family, Vortex-Genie Pulse, Vortex-Genie Recessed Platform