Learn More
Scientific Industries SI™ Foam Inserts for Tubes
Keeps tubes upright and immobile while in use with Vortex-Genie 2. Scientific Industries SI™ Foam Inserts for Tubes fit snugly inside a Vortex-Genie 2 Recessed Platform.
Brand: Scientific Industries SI™ 503-0279-00
Additional Details : Weight : 0.75000kg
Description
- Constructed of high-density foam fitting several 9 tube sizes
- For use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
- One insert is included in the Large Sample Set
Specifications
Tubes | |
High-density foam | |
8 |
14 to 29 mm | |
Vortex-Genie 2 family, Vortex-Genie Pulse, Vortex-Genie Recessed Platform |