Learn More
Foamtec UltraMOP™ III Mops
Bump-Proof™ wrap-around snag-resistant fabric design for disinfecting ISO Class 1 clean room
Brand: Foamtec FS700
Additional Details : Weight : 1.00000kg
Description
- Eliminates rollover and abrasion-related particles
- Enables smooth, skip-free motion, enhancing disinfectant laydown
- Compatible with gamma, autoclave and EtO sterilization methods
Specifications
Foam Mop | |
Sterile | |
Polyurethane Foam |
8.5 in. | |
4.12 in. |