Läs mer
Uvex Fold-flat Mask silv-Air c 3210 FFP2
Handla allt Uvex Produkter
:
Filter Type: FFP2
Beskrivning
General Features
- Particle-filtering folding mask with exhalation valve
- Color: white
Protection Features
- Certified FFP2 face mask to EN 149:2001+A1:2009
- D label (dolomite dust test passed): maximum breathability even with really high dust levels
- Cannot be reused
Comfort Features
- Seamless headband and flexible, adjustable nose clip
- Comfortable sealing lip in nose area for secure fit
- Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
Specifikationer
Specifikationer
| Certifieringar/efterlevnad (Europa) | EN 149 (D marked) |
| Designtyp | Flat-fold |
| Disponibel | Single-use |
| Material | Polypropylene, Polyester |
| Storlek | Standard |
| Med utandningsventil | Yes |
| Filtereffektivitet | FFP2 |
| Sterilitet | Non-sterile |