Beskrivning
General Features
- Particle-filtering flat-fold mask
- Hydrophobic material coating
- Seamless headband for a comfortable, secure fit
- Flexible, adjustable integrated nose clip
- Foam layers in chin and nose area for optimally secure fit
- Individually sealed
- Color: silver
Protection Features
- FFP1 respirator certified in accordance with EN 149:2001+A1:2009
- D marking (mask has passed the dolomite dust test): good breathability even with high levels of dust
- Not reusable
Comfort Features
- Seamless headband for a comfortable, secure fit
- Flexible, adjustable integrated nose clip
- Foam padding on the chin and nose for optimally secure fit
Specifikationer
Specifikationer
| Certifieringar/efterlevnad (Europa) | EN 149 (D marked) |
| Färg | White |
| Designtyp | Flat-fold |
| Disponibel | Single-use |
| Filtertyp | Particulate |
| Material | Polypropylene, Polyester |
| Produkttyp | General Purpose Particulate Respirator |
| Storlek | Standard |
| Med utandningsventil | Yes |
| Filtereffektivitet | FFP1 |
| Visa mer |