Beskrivning
General Features
- Particle-filtering flat-fold mask with climate-optimized exhalation valve
- Hydrophobic material coating
- Four-point headband attachment for a comfortable, individual and secure fit
- Flexible, adjustable integrated nose clip
- Very soft, all-around sealing lip for optimal wearer comfort and secure fit
- Individually sealed
- Color: silver
Protection Features
- FFP3 respirator certified in accordance with EN 149:2001+A1:2009
- D marking (mask has passed the dolomite dust test): good breathability even with high levels of dust
- Not reusable
Comfort Features
- Four-point headband attachment for a comfortable, individual and secure fit
- Flexible, adjustable integrated nose clip
- Very soft, all-around sealing lip for optimal wearer comfort and secure fit
- Exhalation valve to reduce breathing resistance and minimise heat and moisture build-up inside the mask
Specifikationer
Specifikationer
| Aerosol typ | R - Somewhat Resistant to Oil |
| Certifieringar/efterlevnad (Europa) | EN 149 (D marked) |
| Disponibel | Single-use |
| Material | Polypropylene, Polyester |
| Storlek | Standard |
| Med utandningsventil | Yes |
| Filtereffektivitet | FFP3 |
| Sterilitet | Non-sterile |