Beskrivning
General Features
- Individually packed
- Color: white
- With exhalation valve
Safety Features
- Certified FFP2 face mask to EN 149:2001+A1:2009
- D label (dolomite dust test passed): maximum breathability even with really high dust levels
- Cannot be reused
Comfort Features
- Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
- Soft sealing lip in the nose area guarantees a secure, comfortable sealed position of the mask
- Adjustable head band offering easy handling and comfortable fit
- Flexible and adjustable nose clip
- Soft material edges for high wearing comfort
Specifikationer
Specifikationer
| Certifieringar/efterlevnad (Europa) | EN 149 (D marked) |
| Disponibel | Single-use |
| Material | Polypropylene, Polyester |
| Med utandningsventil | Yes |
| Filtereffektivitet | FFP2 |
| Sterilitet | Non-sterile |