Läs mer
Folic Acid-13C5, TRC
Produktkod. 30009424
Change view
Kvantitet:
0.5 mg
5 mg
Förpackningsstorlek:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30009424 5 mg -
30009815 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009424 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF6803035MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1207282-75-4
Molekylformel 13C5 C14 H19 N7 O6
Molekylvikt (g/mol) 446.36
Synonym L-Glutamic-1,2,3,4,5-13C5 acid, N-[4-[[(2-amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl]-, N-[4-[[(2-Amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl]-L-glutamic-1,2,3,4,5-13C5 acid, Folic acid-13C5, Folic acid 13C5 (glutamic acid 13C5), Vitamin B9 13C5
IUPAC-namn (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino](1,2,3,4,5-^{13}C_{5})pentanedioic acid
LEDER NC1=NC(=O)c2nc(CNc3ccc(cc3)C(=O)N[13C@@H]([13CH2][13CH2][13C](=O)O)[13C](=O)O)cnc2N1
Specifikationer
Kemiskt namn eller material Folic acid-13C5
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C19H19N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,8,12,21H,5-7H2,(H,24,29)(H,27,28)(H,31,32)(H3,20,22,25,26,30)/t12-/m0/s1/i5+1,6+1,12+1,13+1,18+1
Formel vikt 446.1565
Procent renhet >95%