missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Follistatin Mouse anti-Human, Mouse, Clone: 4E9B10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60060-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
Follistatin is a single-chain gonadal protein that specifically inhibits follicle-stimulating hormone release. The single FST gene encodes two isoforms, FST317 and FST344 containing 317 and 344 amino acids respectively, resulting from alternative splicing of the precursor mRNA. In a study in which 37 candidate genes were tested for linkage and association with polycystic ovary syndrome (PCOS) or hyperandrogenemia in 150 families, evidence was found for linkage between PCOS and follistatin.Specifications
Follistatin | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P19883, P47931 | |
FST | |
Follistatin Fusion Protein Ag0775 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
4E9B10 | |
Unconjugated | |
FST | |
Activin binding protein, follistatin, FS, FST | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
10468, 14313 | |
-20°C | |
Liquid |