Läs mer
Forasartan, TRC
Produktkod. 30408545
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30408545 1 mg
30395394 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30408545 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF6850001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Forasartan
CAS 145216-43-9
Molekylformel C23 H28 N8
Kvantitet 1 mg
Synonym 5-[(3,5-Dibutyl-1H-1,2,4-triazol-1-yl)methyl]-2-[2-(2H-tetrazol-5-yl)phenyl]pyridine, SC 52458
InChI formel InChI=1S/C23H28N8/c1-3-5-11-21-25-22(12-6-4-2)31(28-21)16-17-13-14-20(24-15-17)18-9-7-8-10-19(18)23-26-29-30-27-23/h7-10,13-15H,3-6,11-12,16H2,1-2H3,(H,26,27,29,30)
LEDER CCCCc1nc(CCCC)n(Cc2ccc(nc2)c3ccccc3c4nnn[nH]4)n1
IUPAC-namn 5-[(3,5-dibutyl-1,2,4-triazol-1-yl)methyl]-2-[2-(1H-tetrazol-5-yl)phenyl]pyridine
Molekylvikt (g/mol) 416.522
Formel vikt 416.244