missing translation for 'onlineSavingsMsg'
Learn More

Formamidine acetate, 99%, Thermo Scientific Chemicals

Catalog Number p-7081261
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
3473-63-0
C3H8N2O2
104.109
MFCD00012866
XPOLVIIHTDKJRY-UHFFFAOYSA-N
formamidine acetate, methanimidamide, monoacetate, formamidinium acetate, acetic acid; methanimidamide, formamidine acetate salt, formamidine, monoacetate, formimidamide acetate, imidoformamide acetate, acetic acid; formamidine, methanimidamide monoacetate
160693
acetic acid;methanimidamide
CC(=O)O.C(=N)N
This item is not returnable. View return policy
3473-63-0
C3H8N2O2
104.109
MFCD00012866
XPOLVIIHTDKJRY-UHFFFAOYSA-N
formamidine acetate, methanimidamide, monoacetate, formamidinium acetate, acetic acid; methanimidamide, formamidine acetate salt, formamidine, monoacetate, formimidamide acetate, imidoformamide acetate, acetic acid; formamidine, methanimidamide monoacetate
160693
acetic acid;methanimidamide
CC(=O)O.C(=N)N

Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.

Solubility
Soluble in water and hot alcohol.

Notes
Hygroscopic. Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼158°C (decomposition)
Density 1.124
pH 8
Quantity 100 g
Beilstein 3563488
Sensitivity Hygroscopic
Solubility Information Soluble in water and hot alcohol.
Formula Weight 104.11
Percent Purity 99%
Chemical Name or Material Formamidine acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'