Läs mer
Formoterol Fumarate Dihydrate, TRC
Produktkod. 30041221
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30041221 25 mg
30041218 100 mg
30040817 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30041221 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF69340025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 183814-30-4
Molekylformel 2C19H24N2O4.C4H4O4.2H2O
Molekylvikt (g/mol) 840.91
Synonym Formoterol Fumarate Dihydrate, Formamide, N-[2-hydroxy-5-[(1R)-1-hydroxy-2-[[(1R)-2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl]-, rel-, (2E)-2-butenedioate (2:1), hydrate (1:2), Formamide, N-[2-hydroxy-5-[(1R)-1-hydroxy-2-[[(1R)-2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl]-, rel-, (2E)-2-butenedioate (2:1) (salt), dihydrate (9CI), Formamide, N-[2-hydroxy-5-[1-hydroxy-2-[[2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl]-, (R*,R*)-, (E)-2-butenedioate (2:1) (salt), dihydrate
IUPAC-namn (E)-but-2-enedioic acid;N-[2-hydroxy-5-[(1R)-1-hydroxy-2-[[(2R)-1-(4-methoxyphenyl)propan-2-yl]amino]ethyl]phenyl]formamide;dihydrate
LEDER O.O.COc1ccc(C[C@@H](C)NC[C@H](O)c2ccc(O)c(NC=O)c2)cc1.COc3ccc(C[C@@H](C)NC[C@H](O)c4ccc(O)c(NC=O)c4)cc3.OC(=O)\C=C\C(=O)O
Specifikationer
Kemiskt namn eller material Formoterol fumarate dihydrate
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/2C19H24N2O4.C4H4O4.2H2O/c2*1-13(9-14-3-6-16(25-2)7-4-14)20-11-19(24)15-5-8-18(23)17(10-15)21-12-22;5-3(6)1-2-4(7)8;;/h2*3-8,10,12-13,19-20,23-24H,9,11H2,1-2H3,(H,21,22);1-2H,(H,5,6)(H,7,8);2*1H2/b;;2-1+;;/t2*13-,19+;;;/m11.../s1
Formel vikt 840.3793
Procent renhet >95%