Läs mer
Formoterol Fumarate(Mixture of Diastereomers), TRC
Produktkod. 30133200
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133200 100 mg
30007699 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133200 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF693401100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 169375-55-7
Molekylformel C19H24N2O4 • C4H4O4
Molekylvikt (g/mol) 460.48
Synonym N-[2-Hydroxy-5-[1-hydroxy-2-[[2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl]-formamide (2E)-2-Butenedioate Salt, Formoterol Hemifumarate
IUPAC-namn N-(2-hydroxy-5-(1-hydroxy-2-((1-(4-methoxyphenyl)propan-2-yl)amino)ethyl)phenyl)formamide fumaric acid
LEDER OC(CNC(C)CC1=CC=C(OC)C=C1)C2=CC=C(O)C(NC=O)=C2.O=C(/C=C/C(O)=O)O
Specifikationer
Kemiskt namn eller material Formoterol Fumarate (Mixture of Diastereomers)
Kvantitet 100 mg
InChI formel InChI=1S/C19H24N2O4.C4H4O4/c1-13(9-14-3-6-16(25-2)7-4-14)20-11-19(24)15-5-8-18(23)17(10-15)21-12-22;5-3(6)1-2-4(7)8/h3-8,10,12-13,19-20,23-24H,9,11H2,1-2H3,(H,21,22);1-2H,(H,5,6)(H,7,8)/b;2-1+
Formel vikt 460.18