Läs mer
Formyl Ciprofloxacin, TRC
Produktkod. 30136586
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136586 25 mg
30135144 5 mg
30007051 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136586 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF69895025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 93594-39-9
Molekylformel C18 H18 F N3 O4
Molekylvikt (g/mol) 359.35
Synonym 1-Cyclopropyl-6-fluoro-7-(4-formyl-1-piperazinyl)-1,4-dihydro-4-oxo-3-quinolinecarboxylic Acid, BAY-p 9357, N-Formylciprofloxacin
IUPAC-namn 1-cyclopropyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
LEDER OC(=O)C1=CN(C2CC2)c3cc(N4CCN(CC4)C=O)c(F)cc3C1=O
Specifikationer
Kemiskt namn eller material Formyl Ciprofloxacin
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C18H18FN3O4/c19-14-7-12-15(8-16(14)21-5-3-20(10-23)4-6-21)22(11-1-2-11)9-13(17(12)24)18(25)26/h7-11H,1-6H2,(H,25,26)
Formel vikt 359.1281
Procent renhet >95%