Learn More
Poulten Graf™ Fortuna™ Class A Glass Blue Color Graduation Pipettes
Provides volumetric accuracy for your pipetting needs. Poulten Graf™ Fortuna™ Class A Glass Blue Color Graduation Pipettes are manufactured from clear glass with permanent blue graduations.
Brand:  Poulten Graf™ 1 1424702
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10140041
  • 809.00 SEK / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
Description
Description
  • Resistant against acids and bases
  • Accuracy better than Class B
  • Pipette aid not necessary
Specifications
Specifications
A
Yes
1 Pack
50 mL
Glass
0.2 mL
Clear
Graduated Pipet