Learn More
Poulten & Graf GmbH FORTUNA™ OPTIMA™ Glass and Metal Syringe with Luer-Lock Tip
Poulten & Graf GmbH FORTUNA™ OPTIMA™ Glass and Metal Syringe with Luer-Lock Tip is used to measure and dispense fluids.
Brand: Poulten & Graf GmbH 7 24044
Additional Details : Weight : 0.10000kg
Description
- With Luer-lock tip
- With steel piston
- Interchangable
- For technical use only
Specifications
Autoclavable | |
Clear | |
Yes | |
No | |
Glass and Metal Syringe | |
30 mL | |
Yes |
Neutral Glass, Neutral Glass (Barrel), Nickel-plated Brass (Tip), Steel (Plunger) | |
Glass and Metal Syringe with Luer-Lock tip | |
Glass and Metal Syringe | |
Amber | |
1 | |
Luer Lock |