Läs mer
Fosamprenavir, MedChemExpress
Produktkod. 16480678
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
Förpackningsstorlek:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16480678 5 mg 5mg
16421989 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16480678 Leverantör Medchem Express Leverantörsnummer HY78726
MedChemExpress Fosamprenavir (Amprenavir phosphate;GW 433908) is a phosphate ester prodrug of the antiretroviral protease inhibitor Amprenavir, with improved solubility. Anti-HIV infection.
Fosamprenavir (Amprenavir phosphate
Specifikationer
Kemiskt namn eller material Fosamprenavir
CAS 226700-79-4
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C25H36N3O9PS
Kvantitet 5 mg
Synonym Amprenavir phosphate GW 433908
Löslighetsinformation DMSO : ≥ 100 mg/mL (170.76 mM)
LEDER O=C(O[C@@H]1COCC1)N[C@@H](CC2=CC=CC=C2)[C@H](OP(O)(O)=O)CN(S(=O)(C3=CC=C(N)C=C3)=O)CC(C)C
Molekylvikt (g/mol) 585.61
Formel vikt 585.61
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-anti-virus