missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOSL1 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB30080.100uL
Additional Details : Weight : 0.02000kg
Description
Rabbit polyclonal antibody raised against synthetic peptide of human FOSL1.
- High specificity
- High reactivity
Specifications
FOSL1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of human FOSL1. | |
In PBS (2% sucrose, 0.09% sodium azide). | |
FRA/FRA1/fra-1 | |
Rabbit | |
Affinity purification | |
RUO | |
8061 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user. | |
NM_005438 | |
FOSL1 | |
A synthetic peptide corresponding to internal region of human FOSL1. | |
100 μL | |
Primary | |
Human | |
Liquid |
For Research Use Only (RUO)