Läs mer
Fosnetupitant, TRC
Produktkod. 30420671
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420671 100 mg
30412570 50 mg
30419521 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30420671 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP831730100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1703748-89-3
Molekylformel C31H35F6N4O5P
Molekylvikt (g/mol) 688.6
Synonym 07PNET, {4-[5-{2-[3,5-bis(trifluoromethyl)phenyl]-N,2-dimethylpropanamido}-4-(2-methylphenyl)pyridin-2-yl]-1-methylpiperazin-1-ium-1-yl}methyl hydrogen phosphate
IUPAC-namn [4-[5-[[2-[3,5-bis(trifluoromethyl)phenyl]-2-methylpropanoyl]-methylamino]-4-(2-methylphenyl)pyridin-2-yl]-1-methylpiperazin-1-ium-1-yl]methyl hydrogen phosphate
LEDER CN(C(=O)C(C)(C)c1cc(cc(c1)C(F)(F)F)C(F)(F)F)c2cnc(cc2c3ccccc3C)N4CC[N+](C)(COP(=O)(O)[O-])CC4
Specifikationer
Kemiskt namn eller material Fosnetupitant
Kvantitet 100 mg
InChI formel InChI=1S/C31H35F6N4O5P/c1-20-8-6-7-9-24(20)25-17-27(40-10-12-41(5,13-11-40)19-46-47(43,44)45)38-18-26(25)39(4)28(42)29(2,3)21-14-22(30(32,33)34)16-23(15-21)31(35,36)37/h6-9,14-18H,10-13,19H2,1-5H3,(H-,43,44,45)
Formel vikt 688.2249