Läs mer
Fosthietan, TRC
Produktkod. 30133737
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133737 250 mg
18012040 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133737 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF730000250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 21548-32-3
Molekylformel C6 H12 N O3 P S2
Molekylvikt (g/mol) 241.27
Synonym Imidocarbonic acid, phosphonodithio-, cyclic methylene P,P-diethyl ester (8CI), Phosphoramidic acid, 1,3-dithietan-2-ylidene-, diethyl ester (9CI), 1,3-Dithietane, phosphoramidic acid deriv., AC 64475, CL 64475, Fosthietan, Geofos, Nematak
IUPAC-namn N-diethoxyphosphoryl-1,3-dithietan-2-imine
LEDER CCOP(=O)(OCC)N=C1SCS1
Specifikationer
Kemiskt namn eller material Fosthietan
Kvantitet 250 mg
InChI formel InChI=1S/C6H12NO3PS2/c1-3-9-11(8,10-4-2)7-6-12-5-13-6/h3-5H2,1-2H3
Formel vikt 240.9996