missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20411-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.Specifications
FOXA1 | |
Polyclonal | |
Unconjugated | |
FOXA1 | |
forkhead box A1, Forkhead box protein A1, FOXA1, HNF 3 alpha, HNF 3A, HNF3A, TCF 3A, TCF3A, Transcription factor 3A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15375, 3169 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35582, P55317 | |
FOXA1 | |
FOXA1 Fusion Protein Ag14243 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |