missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXN1 Mouse anti-Human, Clone: 1D8B12, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | FOXN1 |
---|---|
Clone | 1D8B12 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16898115
|
Proteintech
66337-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16888115
|
Proteintech
66337-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FOXN1 is a 648 amino acid protein belonging to the winged helix/forkhead family It contains 1 fork-head DNA-binding domain and is a transcriptional regulator involved in development. FOXN1 is involved in directing lineage choices of multi-potential progenitor epithelial cells and controls important regulating genes of lympho-epithelial interactions. It modulates keratinocyte differentiation in hair follicles and nail apparatus and its defects lead to T-cell immunodeficiency congenital alopecia, nail dystrophy, macroscopic nudity and an inborn dysgenesis of the thymus.Specifications
FOXN1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FKHL20, forkhead box N1, Forkhead box protein N1, FOXN1, RONU, WHN | |
FOXN1 | |
IgG1 | |
Protein G | |
Antibody |
1D8B12 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O15353 | |
8456 | |
FOXN1 Fusion Protein Ag18207 | |
Primary | |
-20°C | |
FOXN1 |