missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ FOXQ1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518178
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody is predicted to react with mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
FOXQ1 is a member of the FOX gene family, which is characterized by a conserved 110-amino acid DNA-binding motif called the forkhead or winged helix domain. FOX genes are involved in embryonic development, cell cycle regulation, tissue-specific gene expression, cell signaling, and tumorigenesis.
Specifikationer
| FOXQ1 | |
| Polyclonal | |
| Unconjugated | |
| FOXQ1 | |
| forkhead box protein Q1; forkhead box Q1; FOXQ1; Hepatocyte nuclear factor 3 forkhead homolog 1; Hfh1; HFH-1; Hfh1l; HFH-1l; HNF-3/forkhead homolog 1 like; HNF-3/forkhead homolog-1; HNF-3/forkhead-like protein 1; sa; satin; winged helix/forkhead transcription factor | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 94234 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9C009 | |
| FOXQ1 | |
| Synthetic peptide sequence (KLEVFVPRAAHGD) corresponding to the N-terminus amino acids of FOXQ1 (aa 2-15). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering