missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FR 122047 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective cyclooxygenase-1 (COX-1) inhibitor (IC50 values are 0.028 and 65 μM for COX-1 and COX-2 respectively). Antiplatelet, analgesic and anti-inflammatory following oral administration in vivo.
Specifications
Quantity | 10 mg |
Formula Weight | 459.99 |
Percent Purity | >98% |
Chemical Name or Material | FR 122047 hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'