missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FRA7G(D7S486)/CEN7q FISH Probe
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Specifications
Specifications
Content And Storage | Store at 4°C in the dark |
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | fragile site, aphidicolin type, common, fra(7)(q31.2) |
Gene Alias | D7S486 |
Gene Symbol | FRA7G |
Gene ID (Entrez) | 2414 |
Product Type | DNA/RNA |
Quantity | 200 μL |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ FRA7G(D7S486)/CEN7q FISH Probe >
Spot an opportunity for improvement?Share a Content Correction