missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled 10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18175-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
FZD10/Frizzled-10 is a 581 amino acid protein belonging to the G-protein coupled receptor Fz/Smo family and containing a signal peptide, a cysteine-rich domain in the N-terminal, seven transmembrane domains and a C-terminal PDZ domain-binding motif. It is involved in transduction, intercellular transmission of polarity information during tissue morphogenesis in differentiated tissues and as a receptor for Wnt proteins. FZD10 is expressed highly in placenta, fetal kidney, fetal lung, brain cerebellum, cerebral cortex, medulla and spinal cord with very low levels in total brain, frontal lobe, temporal lobe, putamen, adult brain, heart, lung and skeletal muscle.Specifications
Frizzled 10 | |
Polyclonal | |
Unconjugated | |
FZD10 | |
CD350, Frizzled 10, FZ 10, FZD10, FzE7, hFz10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11211, 363913, 93897 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BKG4, Q9ULW2 | |
Fzd10 | |
Frizzled 10 Fusion Protein Ag12867 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |