missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRS2 Mouse anti-Human, Clone: 4E10H12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66263-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
Adapter protein that links activated FGR and NGF receptors to downstream signaling pathways. Plays an important role in the activation of MAP kinases and in the phosphorylation of PIK3R1, the regulatory subunit of phosphatidylinositol 3- kinase, in response to ligand-mediated activation of FGFR1. Modulates signaling via SHC1 by competing for a common binding site on NTRK1.Specifications
FRS2 | |
Monoclonal | |
1.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8WU20 | |
FRS2 | |
FRS2 Fusion Protein Ag19299 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG3 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
4E10H12 | |
Unconjugated | |
FRS2 | |
FGFR signaling adaptor SNT, FGFR substrate 2, FRS2, FRS2A, FRS2alpha, SNT, SNT 1, SNT1 | |
Mouse | |
Protein A | |
RUO | |
10818 | |
-20°C | |
Liquid |