Läs mer
Fructosazine, TRC
Produktkod. 30010190
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010190 10 mg
30012509 25 mg
30004403 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010190 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF78500010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13185-73-4
Molekylformel C12 H20 N2 O8
Molekylvikt (g/mol) 320.3
Synonym (1R,1'R,2S,2'S,3R,3'R)-1,1'-(2,5-Pyrazinediyl)bis-1,2,3,4-butanetetrol;
IUPAC-namn (1R,2S,3R)-1-[5-[(1R,2S,3R)-1,2,3,4-tetrahydroxybutyl]pyrazin-2-yl]butane-1,2,3,4-tetrol
LEDER OC[C@@H](O)[C@@H](O)[C@H](O)c1cnc(cn1)[C@@H](O)[C@H](O)[C@H](O)CO
Specifikationer
Kemiskt namn eller material Fructosazine
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C12H20N2O8/c15-3-7(17)11(21)9(19)5-1-13-6(2-14-5)10(20)12(22)8(18)4-16/h1-2,7-12,15-22H,3-4H2/t7-,8-,9-,10-,11-,12-/m1/s1
Formel vikt 320.122
Procent renhet >95%