missing translation for 'onlineSavingsMsg'
Learn More

Fructose-phenylalanine (mixture of diastereomers), TRC

Product Code. 17948774
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
17948774 10 mg
17939164 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 17948774 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCF79256010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31105-03-0
Molekylformel C15H21NO7
Molekylvikt (g/mol) 327.33
Synonym N-(1-Deoxy-D-fructos-1-yl)-L-phenylalanine, (S)-1-[(1-Carboxy-2-phenylethyl)amino]-1-deoxy-D-fructose, 1-[(α-Carboxyphenethyl)amino]-1-deoxy-fructose, N-(1'-Carboxy-2'-phenylethyl)amino-1-deoxyfructose;
LEDER OC1(CN[C@H](C(O)=O)CC2=CC=CC=C2)[C@@H](O)[C@H](O)[C@H](O)CO1

Specifications

Kemiskt namn eller material Fructose-phenylalanine (mixture of diastereomers)
InChI formel InChI=1S/C15H21NO7/c17-8-12(19)14(21)13(20)11(18)7-16-10(15(22)23)6-9-4-2-1-3-5-9/h1-5,10,12-14,16-17,19-21H,6-8H2,(H,22,23)/t10-,12+,13+,14+/m0/s1
Kvantitet 10 mg
Frakt skick Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.