Läs mer
Fructose-proline (mixture of diastereomers), TRC
Produktkod. 30003258
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003258 10 mg
30006145 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30003258 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF79256510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 29118-61-4
Molekylformel C11H19NO7
Molekylvikt (g/mol) 277.27
Synonym 1-(1-Deoxy-D-fructos-1-yl)-L-proline, (S)-1-(2-Carboxy-1-pyrrolidinyl)-1-deoxy-D-fructose, 1-Deoxy-1-L-proline-D-fructose, 1-Deoxy-1-L-prolino-D-fructose;
LEDER C1C[C@H](N(C1)CC(=O)[C@H]([C@@H]([C@@H](CO)O)O)O)C(=O)O
Specifikationer
Kemiskt namn eller material Fructose-proline (mixture of diastereomers)
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C11H19NO7/c13-5-8(15)10(17)9(16)7(14)4-12-3-1-2-6(12)11(18)19/h6,8-10,13,15-17H,1-5H2,(H,18,19)/t6-,8+,9+,10+/m0/s1
Kvantitet 10 mg
Formel vikt 277.1162
Procent renhet >95%