missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSTL1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | FSTL1 |
---|---|
Concentration | 0.35 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16839694
|
Proteintech
20182-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16829694
|
Proteintech
20182-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein with similarity to follistatin, an activin-binding protein. It contains an FS module, a follistatin-like sequence containing 10 conserved cysteine residues. This gene product is thought to be an autoantigen associated with rheumatoid arthritis.Specifications
FSTL1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat | |
Q12841, Q62632 | |
11167, 79210 | |
FSTL1 Fusion Protein Ag14057 | |
Primary | |
-20°C | |
FSTL1 |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
follistatin like 1, Follistatin like protein 1, Follistatin related protein 1, FRP, FSL1, FSTL1, TSC-36 | |
Fstl1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |