Läs mer
FTBMT, MedChemExpress
Produktkod. 16499976
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
Förpackningsstorlek:
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16499976 50 mg -
16483568 10 mg -
16472776 25 mg -
16426297 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16499976 Leverantör Medchem Express Leverantörsnummer HY101787
MedChemExpress FTBMT is a selective GPR52 agonist with an EC50 of 75 nM. FTBMT has antipsychotic and procognitive properties.
FTBMT is a selective GPR52 agonist with an EC50 of 75 nM. FTBMT has antipsychotic and procognitive properties.
Specifikationer
Kemiskt namn eller material FTBMT
CAS 1358575-02-6
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C19H16F4N4O
Kvantitet 50 mg
Löslighetsinformation DMSO : 50 mg/mL (127.44 mM; Need ultrasonic)
LEDER O=C(N)C1=CC=C(N2C(C)=NC(CC3=CC(C(F)(F)F)=CC(F)=C3)=N2)C=C1C
Molekylvikt (g/mol) 392.35
Formel vikt 392.35
Procent renhet 98.33%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation