missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUBP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10623-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
May interact with single-stranded DNA from the far-upstream element (FUSE). May activate gene expression.Specifications
FUBP3 | |
Polyclonal | |
Unconjugated | |
FUBP3 | |
FBP3, FUBP3, FUSE binding protein 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
320267, 362106, 8939 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96I24 | |
Fubp3 | |
FUBP3 Fusion Protein Ag0974 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |