missing translation for 'onlineSavingsMsg'
Learn More

FUCA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16420-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16886034

  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Alpha-L-fucosidase (EC 3.2.1.51) is a lysosomal enzyme involved in the degradation of fucose-containing glycoproteins and glycolipids.
Specifications

Specifications

FUCA1
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P04066, P17164, Q99LJ1
Alpha L fucosidase 1, Alpha L fucosidase I, FUCA, FUCA1, fucosidase, alpha L 1, tissue, Tissue alpha L fucosidase
Rabbit
IgG
20 μL
-20°C
Primary
24375, 2517, 71665
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FUCA1
Liquid
Fuca1
FUCA1 Fusion Protein Ag9612
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers