Läs mer
Fulvestrant 17-Ketone, TRC
Produktkod. 30006050
Change view
Kvantitet:
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30006050 10 mg -
30004728 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30006050 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF86253510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 403656-89-3
Molekylformel C32H45F5O3S
Molekylvikt (g/mol) 604.75
Synonym (7α)-3-Hydroxy-7-[9-[(4,4,5,5,5-pentafluoropentyl)sulfinyl]nonyl]estra-1,3,5(10)-trien-17-one;
IUPAC-namn (7R,8R,9S,13S,14S)-3-hydroxy-13-methyl-7-[9-(4,4,5,5,5-pentafluoropentylsulfinyl)nonyl]-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-one
LEDER OC1=CC(C[C@@H](CCCCCCCCCS(CCCC(F)(F)C(F)(F)F)=O)[C@]2([H])[C@]3([H])CC[C@@]4(C)[C@@]2([H])CCC4=O)=C3C=C1
Specifikationer
Kemiskt namn eller material Fulvestrant 17-Ketone
Kvantitet 10 mg
InChI formel InChI=1S/C32H45F5O3S/c1-30-17-15-26-25-12-11-24(38)21-23(25)20-22(29(26)27(30)13-14-28(30)39)10-7-5-3-2-4-6-8-18-41(40)19-9-16-31(33,34)32(35,36)37/h11-12,21-22,26-27,29,38H,2-10,13-20H2,1H3/t22-,26-,27+,29-,30+,41?/m1/s1
Formel vikt 604.301