Läs mer
Fumagillin, TRC
Produktkod. 30007321
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007321 10 mg
30004671 5 mg
30003562 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007321 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF86265010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23110-15-8
Molekylformel C26 H34 O7
Molekylvikt (g/mol) 458.54
Synonym 2,4,6,8-Decatetraenedioic acid, 1-[(3R,4S,5S,6R)-5-methoxy-4-[(2R,3R)-2-methyl-3-(3-methyl-2-buten-1-yl)-2-oxiranyl]-1-oxaspiro[2.5]oct-6-yl] ester, (2E,4E,6E,8E)-, 2,4,6,8-Decatetraenedioic acid 4-(1,2-epoxy-1,5-dimethyl-4-hexenyl)-5-methoxy-1-oxaspiro[2.5]oct-6-yl ester (6CI), 2,4,6,8-Decatetraenedioic acid, mono[(3R,4S,5S,6R)-5-methoxy-4-[(2R,3R)-2-methyl-3-(3-methyl-2-butenyl)oxiranyl]-1-oxaspiro[2.5]oct-6-yl] ester, (2E,4E,6E,8E)- (9CI), 2,4,6,8-Decatetraenedioic acid, mono[4-(1,2-epoxy-1,5-dimethyl-4-hexenyl)-5-methoxy-1-oxaspiro[2.5]oct-6-yl] ester (8CI), 2,4,6,8-Decatetraenedioic acid, mono[5-methoxy-4-[2-methyl-3-(3-methyl-2-butenyl)oxiranyl]-1-oxaspiro[2.5]oct-6-yl] ester, [3R-[3α,4α(2R*,3R*),5β,6β(2E,4E,6E,8E)]]-, 1-Oxaspiro[2.5]octane, 2,4,6,8-decatetraenedioic acid deriv., (-)-Fumagillin, Amebacilin, Fugillin, Fumadil B, Fumagillin, Fumagillin DCH, Fumidil, NSC 9168, ZGN 201
IUPAC-namn (2E,4E,6E,8E)-10-[[(3R,4S,5S,6R)-5-methoxy-4-[(2R,3R)-2-methyl-3-(3-methylbut-2-enyl)oxiran-2-yl]-1-oxaspiro[2.5]octan-6-yl]oxy]-10-oxodeca-2,4,6,8-tetraenoic acid
LEDER CO[C@@H]1[C@@H](CC[C@]2(CO2)[C@H]1[C@@]3(C)O[C@@H]3CC=C(C)C)OC(=O)\C=C\C=C\C=C\C=C\C(=O)O
Specifikationer
Kemiskt namn eller material Fumagillin
Kvantitet 10 mg
InChI formel InChI=1S/C26H34O7/c1-18(2)13-14-20-25(3,33-20)24-23(30-4)19(15-16-26(24)17-31-26)32-22(29)12-10-8-6-5-7-9-11-21(27)28/h5-13,19-20,23-24H,14-17H2,1-4H3,(H,27,28)/b7-5+,8-6+,11-9+,12-10+/t19-,20-,23-,24-,25+,26+/m1/s1
Formel vikt 458.2305