Läs mer
Fumaric Acid-13C4, TRC
Produktkod. 30007857
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007857 10 mg
30004625 1 mg
30005249 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007857 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF50038210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 201595-62-2
Molekylformel 13C4H4O4
Molekylvikt (g/mol) 120.04
Synonym (2E)-2-Butenedioic Acid-13C4, (2E)-But-2-enedioic Acid-13C4, (E)-2-Butenedioic Acid-13C4, 2-(E)-Butenedioic Acid-13C4, (E)-Butenedioic Acid-13C4, Allomaleic Acid-13C4, Bakeshure 451-13C4, Bakeshure 470-13C4, Boletic Acid-13C4, FC 33-13C4, Lichenic Acid-13C4, NSC 2752-13C4, Petrom-13C4, trans-1,2-Ethylenedicarboxylic Acid-13C4, trans-2-Butenedioic Acid-13C4, trans-Butenedioic Acid-13C4;
LEDER O=[13C](O)/[13C]([H])=[13C]([H])/[13C](O)=O
Specifikationer
Kemiskt namn eller material Fumaric Acid-13C4
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+/i1+1,2+1,3+1,4+1
Formel vikt 120.0244
Procent renhet >95%