Learn More
Reagecon™ Funhsin Solution Decolorized
Brand: Reagecon™ 1039401
Additional Details : Weight : 0.01000kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Specifications
100 mL | |
Decolorized | |
Funhsin Solution Decolorized |
Safety and Handling
- Danger
Hazard Category
- Carcinogenicity Category 1B
Hazard Statement
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid 37%, 4,4'-(4-iminocyclohexa-2,5-dienylidenemethylene)dianilinehydrochloride