Läs mer
Furaltadone, TRC
Produktkod. 30343398
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30343398 50 mg
1 options
Produktkod. 30343398 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB49733550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Furaltadone
CAS 139-91-3
Molekylformel C13 H16 N4 O6
Kvantitet 50 mg
Synonym 2-Oxazolidinone, 5-(4-morpholinylmethyl)-3-[[(5-nitro-2-furanyl)methylene]amino]-, 2-Oxazolidinone, 5-(morpholinomethyl)-3-[(5-nitrofurfurylidene)amino]- (6CI,7CI,8CI), 3-(5-Nitro-2-furfurylideneamino)-5-(4-morpholinomethyl)-2-oxazolidone, 5-(4-Morpholinomethyl)-3-(5-nitro-2-furfurylideneamino)-2-oxazolidinone, 5-(N-Morpholinomethyl)-3-(5-nitrofurfurylideneamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylidenamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)-2-oxazolidinone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)oxazolidone, Altabactina, Altafur, Anauran, F 150, Fur-Novo, Furaltadon, Furaltadone, Furantoina, Furazolin, Furazoline, Furitale, Furlate, Furlidon, Furmethanol, Furmethonol, Furmetonol, Furmetox, Ibifur, Medifuran, N-(5-Nitro-2-furylidene)-3-amino-5-(N'-morpholinomethyl)-2-oxazolidone, NF 260, NSC 42388, Nitraldone, Nitrofurmethone, Nitrofurmeton, Otifuril, Sepsinol, Ultrafur, Unifur, Valsyn
InChI formel InChI=1S/C13H16N4O6/c18-13-16(14-7-10-1-2-12(22-10)17(19)20)9-11(23-13)8-15-3-5-21-6-4-15/h1-2,7,11H,3-6,8-9H2
LEDER [O-][N+](=O)c1oc(C=NN2CC(CN3CCOCC3)OC2=O)cc1
IUPAC-namn 5-(morpholin-4-ylmethyl)-3-[(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one
Molekylvikt (g/mol) 324.29
Formel vikt 324.107