Läs mer
Furaltadone, TRC
Produktkod. 30333005
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30333005 500 mg
30343970 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30333005 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB497335500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Furaltadone
CAS 139-91-3
Molekylformel C13 H16 N4 O6
Kvantitet 500 mg
Synonym 2-Oxazolidinone, 5-(4-morpholinylmethyl)-3-[[(5-nitro-2-furanyl)methylene]amino]-, 2-Oxazolidinone, 5-(morpholinomethyl)-3-[(5-nitrofurfurylidene)amino]- (6CI,7CI,8CI), 3-(5-Nitro-2-furfurylideneamino)-5-(4-morpholinomethyl)-2-oxazolidone, 5-(4-Morpholinomethyl)-3-(5-nitro-2-furfurylideneamino)-2-oxazolidinone, 5-(N-Morpholinomethyl)-3-(5-nitrofurfurylideneamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylidenamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)-2-oxazolidinone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)oxazolidone, Altabactina, Altafur, Anauran, F 150, Fur-Novo, Furaltadon, Furaltadone, Furantoina, Furazolin, Furazoline, Furitale, Furlate, Furlidon, Furmethanol, Furmethonol, Furmetonol, Furmetox, Ibifur, Medifuran, N-(5-Nitro-2-furylidene)-3-amino-5-(N'-morpholinomethyl)-2-oxazolidone, NF 260, NSC 42388, Nitraldone, Nitrofurmethone, Nitrofurmeton, Otifuril, Sepsinol, Ultrafur, Unifur, Valsyn
InChI formel InChI=1S/C13H16N4O6/c18-13-16(14-7-10-1-2-12(22-10)17(19)20)9-11(23-13)8-15-3-5-21-6-4-15/h1-2,7,11H,3-6,8-9H2
LEDER [O-][N+](=O)c1oc(C=NN2CC(CN3CCOCC3)OC2=O)cc1
IUPAC-namn 5-(morpholin-4-ylmethyl)-3-[(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one
Molekylvikt (g/mol) 324.29
Formel vikt 324.107