Läs mer
Furamidine dihydrochloride, Tocris Bioscience™
Selective PRMT1 inhibitor
Brand: Tocris Bioscience 5202/5
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Furamidine dihydrochloride is a selective protein arginine methyltransferase 1 (PRMT1) inhibitor (IC50 = 9.4 μM). Exhibits selectivity over PRMT5, PRMT6 and CARM1 (IC50 values are 166, 283 and >400 μM respectively). Inhibits cell proliferation in leukemia cell lines. Cell-permeable.Specifikationer
| 4,4'-(2,5-Furandiyl)bis-benzenecarboximidamide dihydrochloride | |
| 55368-40-6 | |
| >98% | |
| 5 mg |