Läs mer
Furamidine Dihydrochloride, TRC
Produktkod. 30408000
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30408000 50 mg
1 options
Produktkod. 30408000 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF86360050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Furamidine Dihydrochloride
CAS 55368-40-6
Molekylformel C18H18Cl2N4O
Kvantitet 50 mg
Synonym 4,4'-(2,5-Furandiyl)bisbenzenecarboximidamide Dihydrochloride, 2,5-Bis(4-amidinophenyl)furan Dihydrochloride, 2,5-Bis(4-guanylphenyl)furan Dihydrochloride, WR 199385
InChI formel InChI=1S/C18H16N4O/c19-17(20)13-5-1-11(2-6-13)15-9-10-16(23-15)12-3-7-14(8-4-12)18(21)22/h1-10H,(H3,19,20)(H3,21,22)
LEDER C1=CC(=CC=C1C2=CC=C(O2)C3=CC=C(C=C3)C(=N)N)C(=N)N
Molekylvikt (g/mol) 377.27