missing translation for 'onlineSavingsMsg'
Learn More

Furan-2-acetic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7077729
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2745-26-8
C6H6O3
126.11
VYSRZETUSAOIMP-UHFFFAOYSA-N
2-furan-2-yl acetic acid, 2-furanacetic acid, 2-furylacetic acid, furan-2-acetic acid, furylacetic acid, unii-c2xfe1jto5, 2-furylaceticacid, furan-2-yl-acetic acid, c2xfe1jto5, 2-2-furyl acetic acid
75974
2-(furan-2-yl)acetic acid
C1=COC(=C1)CC(=O)O
This item is not returnable. View return policy
2745-26-8
C6H6O3
126.11
VYSRZETUSAOIMP-UHFFFAOYSA-N
2-furan-2-yl acetic acid, 2-furanacetic acid, 2-furylacetic acid, furan-2-acetic acid, furylacetic acid, unii-c2xfe1jto5, 2-furylaceticacid, furan-2-yl-acetic acid, c2xfe1jto5, 2-2-furyl acetic acid
75974
2-(furan-2-yl)acetic acid
C1=COC(=C1)CC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 64.0°C to 69.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material Furan-2-acetic acid
compliance-icons
Product Identifier
  • Furan-2-acetic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'