Läs mer
Fusarenon X, TRC
Produktkod. 30406689
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30406689 1 mg
1 options
Produktkod. 30406689 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF8653001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fusarenon X
CAS 23255-69-8
Molekylformel C17 H22 O8
Kvantitet 1 mg
Synonym Trichothec-9-en-8-one, 12,13-epoxy-3α,4β,7α,15-tetrahydroxy-, 4-acetate (8CI), (3α,4β,7α)-4-(Acetyloxy)-12,13-epoxy-3,7,15-trihydroxytrichothec-9-en-8-one, Spiro[2,5-methano-1-benzoxepin-10,2'-oxirane], trichothec-9-en-8-one deriv., 3-Acetylnivalenol, 4-Acetylnivalenol, 4β-Acetoxy-3α,7α,15-trihydroxy-12,13-epoxytrichothec-9-en-8-one, Fusarenon X, Fusarenone, Fusarenone X, NSC 197211, Nivalenol monoacetate
InChI formel InChI=1S/C17H22O8/c1-7-4-9-16(5-18,12(22)10(7)20)15(3)13(24-8(2)19)11(21)14(25-9)17(15)6-23-17/h4,9,11-14,18,21-22H,5-6H2,1-3H3/t9-,11-,12?,13-,14-,15-,16-,17+/m0/s1
LEDER CC(=O)O[C@H]1[C@H](O)[C@@H]2O[C@H]3C=C(C)C(=O)C(O)[C@@]3(CO)[C@@]1(C)[C@@]24CO4
Molekylvikt (g/mol) 354.35
Formel vikt 354.1315