missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | FUT1 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Prix | Quantité et disponibilité | |||||
16877974
|
Proteintech
17956-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867974
|
Proteintech
17956-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FUT1 is a Golgi stack membrane protein that is involved in the creation of a precursor of the H antigen, which is required for the final step in the soluble A and B antigen synthesis pathway.Spécification
FUT1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O09160, P19526 | |
14343, 2523 | |
FUT1 Fusion Protein Ag12450 | |
Primary | |
-20°C | |
FUT1 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Alpha(1,2)FT 1, Fucosyltransferase 1, FUT1, H, HH, HSC | |
FUT1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |