missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17175-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Probable fucosyltransferase.Specifications
FUT11 | |
Polyclonal | |
Unconjugated | |
FUT11 | |
Fuc TXI, Fucosyltransferase 11, Fucosyltransferase XI, FucT XI, FUT11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170384, 286971, 73068 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q495W5, Q68FV3, Q8BHC9 | |
Fut11 | |
FUT11 Fusion Protein Ag10815 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |