Learn More
Invitrogen™ FXR Human Protein, Ligand Binding Domain, GST-Tagged
Farnesoid X Receptor, Ligand Binding Domain (FXR-LBD) is a 60.07 kDa recombinant human protein (amino acids 193-472) expressed as a GST fusion protein in baculovirus-infected insect cells.
Brand:  Invitrogen™ PV4835
Additional Details : Weight : 0.50000kg
Product Code. 10668693
  • 17750.00 SEK / Each
Estimated Shipment: 14-05-2024
to see stock.
For Research Use Only. All usage must comply with product instructions.
Description
Description
FXR-LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Specifications
Specifications
Nuclear Receptor Assay
50 μg
RUO - research use only
FXR LBD
Recombinant
9971
Store in ultra-cold freezer (-68 to -85°C).
Ligand Binding Domain
Human
Hormone Receptors