Learn More
Invitrogen™ FXR Human Protein, Ligand Binding Domain, GST-Tagged
Farnesoid X Receptor, Ligand Binding Domain (FXR-LBD) is a 60.07 kDa recombinant human protein (amino acids 193-472) expressed as a GST fusion protein in baculovirus-infected insect cells.
Brand: Invitrogen™ PV4835
Additional Details : Weight : 0.50000kg
Description
FXR-LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Specifications
Nuclear Receptor Assay | |
50 μg | |
RUO - research use only | |
FXR LBD | |
Recombinant |
9971 | |
Store in ultra-cold freezer (-68 to -85°C). | |
Ligand Binding Domain | |
Human | |
Hormone Receptors |