missing translation for 'onlineSavingsMsg'
Learn More

FXR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12552-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16840314

  • 3313.85 SEK / 150µL
Estimated Shipment: 03-10-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Fragile X syndrome is caused by the absence of the fragile X mental-retardation protein (FMRP). FMRP is the archetype of a class of cytoplasmic mRNA-binding proteins that includes the fragile X-related 1 and 2 proteins (FXR1 and FXR2). The fragile X-related proteins FXR1 and FXR2 contain a functional nucleolar-targeting signal equivalent to the HIV-1 regulatory proteins.
Specifications

Specifications

FXR2
Polyclonal
Unconjugated
FXR2
FMR1L2, FXR2
Rabbit
Antigen Affinity Chromatography
RUO
23879, 287433, 9513
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P51116, Q9WVR4
Fxr2
FXR2 Fusion Protein Ag3402
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers