missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FY (Human) Recombinant Protein
Human FY full-length ORF ( AAH17817, 1 a.a. - 336 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002532-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MGNCLHRAELSPSTENSSQLDFEDVWNSSYGVNDSFPDGDYGANLEAAAPCHSCNLLDDSALPFFILTSVLGILASSTVLSMLFRPLFRWQLCPGWPVLAQLAVGSALFSIVVPVLAPGLGSTRSSALCSLGYCVWYGSAFAQALLLGCHASLGHRLGAGQVPGLTLGLTVGIWGVAALLTLPVTLASGASGGLCTLIYSTELKALQATHTVACLAIFVLLPLGLFGAKGLKKALGMGPGPWMNILWAWFIFWWPHGVVLGLDFLVRSKLLLLSTCLAQQALDLLLNLAEALAILHCVATPLLLALFCHQATRTLLPSLPLPEGWFSHLDTLGSKS
Specifications
AAH17817 | |
Duffy blood group, chemokine receptor | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DARC | |
GST |
2532 | |
Wheat germ expression system | |
10 μg | |
CCBP1, CD234, Dfy, FY, GPD, GpFy, WBCQ1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |